Assay Details
| Gene Symbol: SAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598628.2 | XP_005598685.1 | 851 | 120 | ||
| XM_014729883.2 | XP_014585369.1 | 941 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAP25| Gene Symbol: SAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598628.2 | XP_005598685.1 | 851 | 120 | ||
| XM_014729883.2 | XP_014585369.1 | 941 | 120 | |||
Set Membership: |
|