Assay Details
| Gene Symbol: LOC100060748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492921.6 | XP_001492971.2 | 1199 | 73 | ||
| XM_023655407.1 | XP_023511175.1 | 1201 | 73 | |||
More Information
Set Membership: |
|

LOC100060748
Genomic Map