Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615124.2 | XP_005615181.2 | 1399 | 103 | ||
| XM_023655427.1 | XP_023511195.1 | 1318 | 103 | |||
| XM_023655428.1 | XP_023511196.1 | 1084 | 103 | |||
| XM_023655429.1 | XP_023511197.1 | 927 | 103 | |||
More Information
Set Membership: |
|

POM121C
Genomic Map