Assay Details
| Gene Symbol: LOC100062625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494135.4 | XP_001494185.1 | 364 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494135.4 transcripts.
Set Membership: |
|

LOC100062625
Genomic Map