Assay Details
| Gene Symbol: LOC100062625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494135.4 | XP_001494185.1 | 1036 | 66 | ||
| XM_023655502.1 | XP_023511270.1 | 1094 | 66 | |||
| XM_023655503.1 | XP_023511271.1 | 1095 | 66 | |||
| XM_023655504.1 | XP_023511272.1 | 1095 | 66 | |||
More Information
Set Membership: |
|

LOC100062625
Genomic Map