Assay Details
Gene Symbol: LOC100062082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598712.3 | XP_005598769.2 | 1269 | 70 | ||
XM_014729938.2 | XP_014585424.2 | 1320 | 70 | |||
XM_023655521.1 | XP_023511289.1 | 1321 | 70 | |||
XM_023655522.1 | XP_023511290.1 | 1269 | 70 | |||
XM_023655523.1 | XP_023511291.1 | 1127 | 70 | |||
XM_023655524.1 | XP_023511292.1 | 1322 | 70 | |||
XM_023655525.1 | XP_023511293.1 | 1280 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
