Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729955.2 | XP_014585441.1 | 279 | 121 | ||
| XM_014729956.2 | XP_014585442.1 | 225 | 121 | |||
| XM_014729957.2 | XP_014585443.1 | 152 | 121 | |||
| XM_023655563.1 | XP_023511331.1 | 710 | 121 | |||
More Information
Set Membership: |
|

ZNF713
Genomic Map