Assay Details
| Gene Symbol: LOC100064310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730035.2 | XP_014585521.2 | 1499 | 76 | ||
| XM_023616051.1 | XP_023471819.1 | 1496 | 76 | |||
More Information
Set Membership: |
|

LOC100064310
Genomic Map