Assay Details
Gene Symbol: LOC100064310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730035.2 | XP_014585521.2 | 544 | 90 | ||
XM_023616051.1 | XP_023471819.1 | 544 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
