Assay Details
Gene Symbol: LOC100058833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616174.1 | XP_023471942.1 | 2344 | 88 | ||
XM_023616175.1 | XP_023471943.1 | 2138 | 88 | |||
XM_023616177.1 | XP_023471945.1 | 2344 | 88 | |||
XM_023616178.1 | XP_023471946.1 | 2344 | 88 | |||
XM_023616180.1 | XP_023471948.1 | 1966 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
