Assay Details
Gene Symbol: LOC100058987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491761.5 | XP_001491811.3 | 1282 | 77 | ||
XM_023616173.1 | XP_023471941.1 | 1152 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
