Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598931.3 | XP_005598988.3 | 776 | 92 | ||
| XM_005598932.2 | XP_005598989.1 | 700 | 92 | |||
| XM_023616349.1 | XP_023472117.1 | 405 | 92 | |||
More Information
Set Membership: |
|

CARHSP1
Genomic Map