Assay Details
Gene Symbol: SMIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598986.3 | XP_005599043.1 | 282 | 72 | ||
XM_014730164.2 | XP_014585650.1 | 282 | 72 | |||
XM_014730165.2 | XP_014585651.1 | 621 | 72 | |||
XM_014730167.2 | XP_014585653.1 | 501 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
