Assay Details
Gene Symbol: SEPTIN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598989.3 | XP_005599046.2 | 822 | 103 | ||
XM_023616374.1 | XP_023472142.1 | 864 | 103 | |||
XM_023616375.1 | XP_023472143.1 | 837 | 103 | |||
XM_023616376.1 | XP_023472144.1 | 711 | 103 | |||
XM_023616377.1 | XP_023472145.1 | 864 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
