Assay Details
| Gene Symbol: PAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599005.3 | XP_005599062.1 | 147 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005599005.3 transcripts.
Set Membership: |
|

PAM16
Genomic Map