Assay Details
Gene Symbol: LOC106781592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617474.1 | XP_023473242.1 | 294 | 121 | ||
XM_023617475.1 | XP_023473243.1 | 290 | 121 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
