Assay Details
| Gene Symbol: C14H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599410.3 | XP_005599467.1 | 696 | 69 | ||
| XM_023617675.1 | XP_023473443.1 | 697 | 69 | |||
More Information
Set Membership: |
|

C14H5orf24
Genomic Map