Assay Details
Gene Symbol: LOC100061234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627061.1 | XP_023482829.1 | 753 | 125 | ||
XM_023627062.1 | XP_023482830.1 | 909 | 125 | |||
XM_023627063.1 | XP_023482831.1 | 939 | 125 | |||
XM_023627064.1 | XP_023482832.1 | 768 | 125 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
