Assay Details
| Gene Symbol: C21H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604103.3 | XP_005604160.1 | 961 | 88 | ||
| XM_014734757.2 | XP_014590243.1 | 960 | 88 | |||
| XM_014734758.2 | XP_014590244.1 | 958 | 88 | |||
| XM_014734759.2 | XP_014590245.1 | 959 | 88 | |||
| XM_023625371.1 | XP_023481139.1 | 910 | 88 | |||
| XM_023625372.1 | XP_023481140.1 | 958 | 88 | |||

C21H19orf44
Genomic Map