Assay Details
| Gene Symbol: LOC106781693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619721.1 | XP_023475489.1 | 208 | 95 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106781693| Gene Symbol: LOC106781693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619721.1 | XP_023475489.1 | 208 | 95 | ||
Set Membership: | 
		   
					
						
							 |