Assay Details
Gene Symbol: ZC2HC1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491724.4 | XP_001491774.2 | 150 | 106 | ||
XM_005605337.3 | XP_005605394.1 | 149 | 106 | |||
XM_005605340.3 | XP_005605397.1 | 149 | 106 | |||
XM_023628133.1 | XP_023483901.1 | 150 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491724.4, XM_005605337.3, XM_005605340.3, XM_023628133.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
