Assay Details
| Gene Symbol: C23H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627057.1 | XP_023482825.1 | 162 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C23H9orf92| Gene Symbol: C23H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627057.1 | XP_023482825.1 | 162 | 88 | ||