Assay Details
| Gene Symbol: FYB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629082.1 | XP_023484850.1 | 1619 | 94 | ||
| XM_023629091.1 | XP_023484859.1 | 1619 | 94 | |||
| XM_023629101.1 | XP_023484869.1 | 1619 | 94 | |||
More Information
Set Membership: |
|

FYB2
Genomic Map