Assay Details
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633462.1 | XP_023489230.1 | 939 | 92 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633462.1 | XP_023489230.1 | 939 | 92 |