Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634414.1 | XP_023490182.1 | 753 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
