Assay Details
Gene Symbol: TAS2R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639845.1 | XP_023495613.1 | 4321 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
