Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641295.1 | XP_023497063.1 | 921 | 80 | ||
XM_023641296.1 | XP_023497064.1 | 751 | 80 | |||
XM_023641297.1 | XP_023497065.1 | 176 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023641297.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
