Assay Details
| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645647.1 | XP_023501415.1 | 2118 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM181B| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645647.1 | XP_023501415.1 | 2118 | 90 | ||
Set Membership: |
|