Assay Details
| Gene Symbol: RBMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645954.1 | XP_023501722.1 | 5598 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMXL2| Gene Symbol: RBMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645954.1 | XP_023501722.1 | 5598 | 73 | ||