Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649416.1 | XP_023505184.1 | 520 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
