Assay Details
Gene Symbol: LOC102148743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650298.1 | XP_023506066.1 | 885 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
