Assay Details
| Gene Symbol: CCDC92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653245.1 | XP_023509013.1 | 893 | 62 | ||
| XM_023653246.1 | XP_023509014.1 | 383 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC92B| Gene Symbol: CCDC92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653245.1 | XP_023509013.1 | 893 | 62 | ||
| XM_023653246.1 | XP_023509014.1 | 383 | 62 | |||
Set Membership: |
|