Assay Details
Gene Symbol: BHLHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653751.1 | XP_023509519.1 | 825 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
