Assay Details
Gene Symbol: MRGPRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654901.1 | XP_023510669.1 | 140 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
