Assay Details
Gene Symbol: PTPRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497719.5 | XP_001497769.1 | 139 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001497719.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
