Assay Details
Gene Symbol: OR56B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499362.2 | XP_001499412.2 | 916 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
