Assay Details
| Gene Symbol: C7H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504582.5 | XP_001504632.1 | 142 | 97 | ||
| XM_005612113.3 | XP_005612170.1 | 142 | 97 | |||
More Information
Set Membership: |
|

C7H11orf42
Genomic Map