Assay Details
Gene Symbol: LOC100146589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915637.4 | XP_001915672.1 | 715 | 54 | ||
XM_023641017.1 | XP_023496785.1 | 654 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
