Assay Details
| Gene Symbol: FOXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917120.4 | XP_001917155.2 | 323 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001917120.4 transcripts.
Set Membership: |
|

FOXD4
Genomic Map