Assay Details
| Gene Symbol: LOC100056062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123381.1 | NP_001116853.1 | 5 - 6 | 1004 | 92 | |
| XM_023624047.1 | XP_023479815.1 | 1074 | 92 | |||
| GenBank mRNA | DQ145609.1 | - | 4 - 5 | 723 | 92 | |
| EF577071.1 | - | 4 - 5 | 723 | 92 | ||
| KC291655.1 | - | 5 - 6 | 1022 | 92 | ||
More Information
Set Membership: |
|

LOC100056062
Genomic Map