Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252255.1 | NP_001239184.1 | 2 - 3 | 390 | 72 | |
| XM_015293128.2 | XP_015148614.1 | 2 - 3 | 218 | 72 | ||
| GenBank mRNA | BX929563.1 | - | 1 - 2 | 502 | 72 | |
| BX930684.1 | - | 2 - 3 | 390 | 72 | ||
| BX931261.1 | - | 2 - 3 | 303 | 72 | ||
| BX931502.2 | - | 1 - 2 | 507 | 72 | ||
More Information
Set Membership: |
|

RPL32
Genomic Map