Assay Details
| Gene Symbol: PRRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199415.1 | NP_001186344.1 | 5 - 6 | 521 | 91 | |
| XM_015286158.2 | XP_015141644.1 | 4 - 5 | 892 | 91 | ||
| GenBank mRNA | BU301058.1 | - | 5 - 6 | 501 | 91 | |
| BX929762.1 | - | 5 - 6 | 455 | 91 | ||
More Information
Set Membership: |
|

PRRG4
Genomic Map