Assay Details
| Gene Symbol: DHRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277910.3 | NP_001264839.1 | 3 - 4 | 898 | 86 | |
| XM_025142529.1 | XP_024998297.1 | 6 - 7 | 953 | 86 | ||
| XM_025142530.1 | XP_024998298.1 | 8 - 9 | 1111 | 86 | ||
| XM_025142531.1 | XP_024998299.1 | 3 - 4 | 427 | 86 | ||
| XM_025142532.1 | XP_024998300.1 | 3 - 4 | 368 | 86 | ||
| GenBank mRNA | BX929880.1 | - | 3 - 4 | 474 | 86 | |
| BX950638.1 | - | 3 - 4 | 905 | 86 | ||
More Information
Set Membership: |
|

DHRS3
Genomic Map