Assay Details
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285926.1 | NP_001272855.1 | 6 - 7 | 555 | 69 | |
XM_004938247.3 | XP_004938304.1 | 6 - 7 | 639 | 69 | ||
XM_004938248.3 | XP_004938305.1 | 6 - 7 | 555 | 69 | ||
GenBank mRNA | BI065382.1 | - | 5 - 6 | 398 | 69 | |
BU433840.1 | - | 2 - 3 | 282 | 69 | ||
BX930121.1 | - | 6 - 7 | 552 | 69 | ||
BX930142.2 | - | 7 - 8 | 702 | 69 | ||
BX930156.1 | - | 7 - 8 | 702 | 69 | ||
BX930576.2 | - | 6 - 7 | 552 | 69 | ||
BX930906.1 | - | 6 - 7 | 565 | 69 | ||
BX931243.2 | - | 6 - 7 | 552 | 69 | ||
BX933264.1 | - | 6 - 7 | 535 | 69 | ||
BX935375.1 | - | 6 - 7 | 555 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
