Assay Details
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321600.1 | NP_001308529.1 | 5 - 6 | 601 | 130 | |
XM_015282055.2 | XP_015137541.1 | 5 - 6 | 843 | 130 | ||
XM_015282056.2 | XP_015137542.1 | 5 - 6 | 861 | 130 | ||
XM_025147177.1 | XP_025002945.1 | 5 - 6 | 855 | 130 | ||
XM_025147178.1 | XP_025002946.1 | 3 - 4 | 368 | 130 | ||
GenBank mRNA | BX930389.1 | - | 5 - 6 | 504 | 130 | |
BX950353.2 | - | 5 - 6 | 570 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
