Assay Details
Gene Symbol: COX6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198657.1 | NP_001185586.1 | 1 - 2 | 120 | 58 | |
GenBank mRNA | BX930527.1 | - | 1 - 2 | 120 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198657.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
