Assay Details
Gene Symbol: H2AFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277338.2 | NP_001264267.1 | 1 - 2 | 235 | 65 | |
XM_015288210.1 | XP_015143696.1 | 2 - 3 | 734 | 65 | ||
XM_015288211.2 | XP_015143697.1 | 2 - 3 | 380 | 65 | ||
GenBank mRNA | BX930607.1 | - | 2 - 3 | 354 | 65 | |
BX931423.2 | - | 2 - 3 | 281 | 65 |
Gene Symbol: LOC112532616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151724.1 | XP_025007492.1 | 2 - 3 | 378 | 65 | |
XM_025151725.1 | XP_025007493.1 | 2 - 3 | 386 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
