Assay Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278022.1 | NP_001264951.1 | 1 - 2 | 335 | 103 | |
XM_015284633.1 | XP_015140119.1 | 1 - 2 | 3142 | 103 | ||
GenBank mRNA | BX930608.1 | - | 1 - 2 | 335 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
