Assay Details
| Gene Symbol: MSRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199578.1 | NP_001186507.1 | 3 - 4 | 259 | 149 | |
| XM_015281611.2 | XP_015137097.1 | 4 - 5 | 429 | 149 | ||
| XM_015281618.2 | XP_015137104.1 | 3 - 4 | 329 | 149 | ||
| XM_015281622.2 | XP_015137108.2 | 4 - 5 | 401 | 149 | ||
| XM_015281636.2 | XP_015137122.1 | 4 - 5 | 567 | 149 | ||
| XM_015281639.2 | XP_015137125.1 | 4 - 5 | 392 | 149 | ||
| XM_015281645.2 | XP_015137131.1 | 2 - 3 | 232 | 149 | ||
| GenBank mRNA | BX930748.1 | - | 1 - 2 | 220 | 149 | |
| BX933182.1 | - | 3 - 4 | 259 | 149 | ||
More Information
Set Membership: |
|

MSRB3
Genomic Map