Assay Details
| Gene Symbol: PSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424846.6 | XP_424846.3 | 5 - 6 | 619 | 96 | |
| GenBank mRNA | BX929450.1 | - | 5 - 6 | 577 | 96 | |
| BX930805.1 | - | 5 - 6 | 581 | 96 | ||
| BX935484.2 | - | 5 - 6 | 604 | 96 | ||
More Information
Set Membership: |
|

PSAT1
Genomic Map